Affinity Protein Expression and Purification Systems, Agilent Technologies
Catalog #: CAST214303
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
CAS Number:
Specifications
- Description:Calmodulin Affinity Resin
- Size:10 ml
- Cat. no.:CAST214303
- Details:Calmodulin binding peptide (CBP) affinity resin.
Specifications
About this item
The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purification.
- pCAL vectors derived from T7 RNA polymerase-based pET vectors
- Novel purification tag combines small size, gentle elution and high-affinity binding
- Easy tag removal resulting in native protein
Delivery: The complete system includes pCAL expression vectors containing a calmodulin-binding peptide tag and calmodulin affinity resin (available separately).
Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought