Order Entry
ContactUsLinkComponent
GeneMorph II Random Mutagenesis Kit 30 reactions
Random Mutagenesis Kits GeneMorph II, Agilent Technologies
Catalog #: CAST200550
CAS Number:  
GeneMorph II Random Mutagenesis Kit 30 reactions
Random Mutagenesis Kits GeneMorph II, Agilent Technologies
Catalog #: CAST200550
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    GeneMorph II Random Mutagenesis Kit
  • Cat. no.:
    CAST200550
  • No. of Reactions:
    30

Specifications

About this item

The GeneMorph II random mutagenesis kits have been formulated to produce a more uniform mutational spectrum when performing error prone PCR to achieve random mutagenesis.

  • Superior yields over a wide target range - 0.1 to 6 kb
  • Easily change mutation frequency by altering concentration of input template
  • Efficient mutagenesis rates of 1 to 16 bases per kb

Other methods or commercially available kits that rely on Taq DNA polymerase for random mutagenesis have shown biases towards mutating A’s and T’s more frequently than G’s and C’s, which undoubtedly skews representation of random mutant libraries.

GeneMorph II kits utilize Mutazyme II DNA polymerase, which is a novel error prone PCR enzyme blend with equivalent mutation rates at A’s and T’s vs. G’s and C’s. Additionally, to address the need for efficient and flexible cloning methods while delivering this less biased mutational spectrum. The GeneMorph II random mutagenesis kit takes error-prone PCR to the next level by combining mutational spectrums of both our Mutazyme DNA polymerase and Taq DNA polymerase

GeneMorph II EZClone Domain Mutagenesis Kit offers an easy and fast cloning method to perform targeted random mutagenesis on protein domains and promoter elements and eliminates the need to have specific restriction sites.

Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought