QuikChange II, Agilent Technologies
Catalog #: CAST200522
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:QuikChange II XL Site-Directed Mutagenesis Kit
- Cat. no.:CAST200522
- No. of Reactions:30
- Details:The highest fidelity mutagenesis kit for large plasmids
Specifications
About this item
These QuikChange II kits provide improved fidelity, while maintaining greater than 80% mutation efficiency for single site mutagenesis.
- Includes PfuUltra High-fidelity DNA polymerase to minimize unwanted errors
- Complete kit provides all reagents necessary to support mutagenesis (at single sites)
Used for large constructs, and for use with electroporation competent cells.
Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought