ContactUsLinkComponent
Polypropylene 96-Well Tube Plates 25 Plates
PCR and qPCR Consumables, Agilent Technologies
Catalog #: CA99900-152
Polypropylene 96-Well Tube Plates 25 Plates
PCR and qPCR Consumables, Agilent Technologies
Catalog #: CA99900-152
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Polypropylene 96-well tube plates, 25 plates/pack
  • Cat. no.:
    CA99900-152
  • Details:
    96-Well Non-Skirted polypropylene PCR Plates for use with Mx3000P / Mx3005P qPCR Systems
Specifications
About this item
High-quality well plate supplies and accessories will enhance your PCR and qPCR workflow and analysis.
Agilent AriaMx low profile strip tubes are optimized for PCR and qPCR applications. The tubes are validated for optimal performance with Mx3005P, Mx3000P, and the AriaMx real time PCR Systems. Part of the qPCR Plastics and Accessories, the products include the perfect fit frame, tubes, and caps.
Additional products available include the 96-well semiskirted polypropylene PCR plate compatible with the optical Strip Caps, the StrataCoolers LP Benchtop Cooler, and StrataCooler cryo preservation module. Tubes and accessories are compatible most PCR and Real-Time instruments.
Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought