Order Entry
ContactUsLinkComponent
QuikChange Site-Directed Mutagenesis Kit 10 reactions
Mutagenesis Kits QuikChange, Agilent Technologies
Catalog #: CA99900-052
CAS Number:  
QuikChange Site-Directed Mutagenesis Kit 10 reactions
Mutagenesis Kits QuikChange, Agilent Technologies
Catalog #: CA99900-052
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    QuikChange Site-Directed Mutagenesis Kit
  • Cat. no.:
    CA99900-052
  • No. of Reactions:
    10

Specifications

About this item

The original QuikChange Site-Directed Mutagenesis Kits speed up and simplify site-directed mutagenesis studies. The kits eliminate the need for sub-cloning into M13-based bacteriophage vectors and for ss-DNA rescue. This allows oligo-mediated introduction of site-specific mutations into virtually any double-stranded plasmid DNA. In addition, the XL version of the kit is specially designed for efficient mutagenesis of large (4 -14 kb) or otherwise difficult-to mutagenize plasmid templates. The XL kit features components specifically designed for more efficient DNA replication and bacterial transformation.

  • >80% mutagenesis efficiency for one site; >55% mutagenesis efficiency for three mutations simultaneously
  • Efficient and accurate linear amplification minimizes unwanted errors

QuikChange Site-Directed Mutagenesis Kit: This kit uses PfuTurbo DNA polymerase. It replicates plasmid strands with high fidelityll without displacing mutant oligonucleotide primers. The process uses a supercoiled dsDNA vector with insert and two synthetic oligo primers containing the mutation. Incorporating oligo primers creates a mutated plasmid with staggered nicks. The product is treated with Dpn I after temperature cycling. Dpn I digests the parental DNA template, selecting mutation-containing synthesized DNA. The nicked vector DNA containing the mutations is transformed into XL1-Blue supercompetent cells.

QuikChange XL Site-Directed Mutagenesis Kit: The QuikChange II XL site-directed mutagenesis kit specializes in efficient mutagenesis of large (8–14 kb) or otherwise difficult-to mutagenize plasmid templates and features components designed for more efficient DNA replication and bacterial transformation. A QuikSolution reagent is provided to facilitate replication of large plasmids, while XL10-Gold Ultracompetent Cells have been included to ensure the highest transformation efficiencies possible. XL10-Gold cells contain the Hte phenotype, increasing the transformation efficiency of larger DNA plasmids.

QuikChange Multi site-directed mutagenesis kit: The QuikChange Multi site-directed mutagenesis system is a novel technology that allows mutagenesis at multiple sites in a single round, using a single oligonucleotide per site. This system simplifies randomizing key amino acids using oligos containing degenerate codons. No specialized vectors or unique restriction sites are needed to use the kit; virtually any plasmid up to 8 kb is a suitable template. The rapid three-step procedure introduces mutations at three different sites simultaneously in the 4 kb QuikChange Multi control plasmid with >50% efficiency.

Caution: Academic QuikChange Multi site-directed mutagenesis Kits are for Academic Use Only
Customers Who Bought This Also Bought