Order Entry
ContactUsLinkComponent
Brilliant II SYBR Green Low ROX QPCR Master Mix 400 reactions
Brilliant II SYBR® MM with ROX, Agilent Technologies
Catalog #: CA97066-496
CAS Number:  
Brilliant II SYBR Green Low ROX QPCR Master Mix 400 reactions
Brilliant II SYBR® MM with ROX, Agilent Technologies
Catalog #: CA97066-496
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
    qPCR dye
  • Description:
    Brilliant II SYBR Green Low ROX QPCR Master Mix
  • No. of Reactions:
    400
  • Details:
    High performance, ultra sensitive, brighter and faster next generation SYBR(R) Green QPCR master mix reagent with ROX reference dye for reliable quantification across a wide range of targets and templates

Specifications

About this item

Brilliant II SYBR® QPCR and QRT-PCR master mix kits with high/low Rox offer superior sensitivity for improved quantification and reproducibility.

  • Improved performance with earlier Ct detection across a wide dynamic range
  • Increased sensitivity of detection to low copy numbers

It also shows more consistent performance across a wide range of template concentrations and different targets.

Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought