Ward's® Mini Benchtop Coolers
Catalog #: 470306-952
Supplier: Heathrow Scientific
Specifications
- Color:Blue with clear lid
- Description:32 place cooler for 0.5 - 2 ml tubes
- Cat. no.:470306-952
- Dimensions:9.6 L×4.1 W×3.9" H
- Cooling Capacity:−20 °C
Specifications
About this item
Protect temperature-sensitive samples at the bench or in the freezer.
- Polycarbonate construction with rubber non-slip feet
- Gridded and keyed lid with one position for easy sample identification
Ideal for keeping samples or reagents cool during testing.
Customers Who Bought This Also Bought