Lab-TAPE™ Color Lab Tapes, GA International
Catalog #: 76275-742
Supplier: GA INTERNATIONAL INC
Specifications
- Dimensions:0.94" × 180'
- Color:Pink
- Cat. no.:76275-742
Specifications
About this item
Color laboratory tape is available in a wide range of colors and widths, for easy identification and color-coding.
- Ideal for all general use applications in the lab
- Continuous tape format allows use of as much or little as necessary
- Resistant to gamma irradiation (tested to up to 50 kGy)
Available in an assortment of eye-catching colors and various widths, Lab tape is perfect for color-coding and general lab use. Can be written on with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.
Customers Who Bought This Also Bought