Order Entry
ContactUsLinkComponent
Ribbon Resin Bk  0.5In 3.3Inx244Ft RL1
Thermal Transfer Ribbons, GA International
Catalog #: 76274-760
Ribbon Resin Bk  0.5In 3.3Inx244Ft RL1
Thermal Transfer Ribbons, GA International
Catalog #: 76274-760

Specifications

  • Color:
    Black
  • Description:
    Smudge proof resin ribbon
  • Cat. no.:
    76274-760
  • Length:
    74.3 m (244 ft.)
  • Width:
    84 mm (3.3")

Specifications

About this item

Customers Who Bought This Also Bought