Tubing Connectors, CADENCE SCIENCE®
Catalog #: 76269-004
Supplier: Cadence Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Tubing I.D.:⅛ - ³⁄₁₆"
- Description:Female Luer Lock Tubing Connector
- Material:Plated brass
- Cat. no.:76269-004
Specifications
About this item
These tubing connectors are used in hospitals, laboratories, and biomedical instruments. The metal connectors are precision machined and are made from either nickel plated brass or 316 stainless steel.
Customers Who Bought This Also Bought