Erlenmeyer Shaker Flasks, Greiner Bio-One
Catalog #: 76266-256
Supplier: Greiner Bio-One
Specifications
- Capacity:125 ml
- Color:Clear
- Description:Erlenmeyer Shaker Flask
- Cat. no.:76266-256
Specifications
About this item
These erlenmeyer shaker flasks are ideal for suspension culture of mammalian cells, plants, microbes and microorganisms. The shaker flasks feature a patented 2-in-1 DuoCAP® which allows for flexibility between vented and non-vented applications.
- Individually packaged and sterile to SAL 10-6 for long term culture applications
- Molded-in graduations for easy volumetric reference
- Non-pyrogenic, non-cytotoxic, and nuclease-free
Each flask is manufactured utilizing an optically clear, inert polycarbonate resin which can be disposed of after single use or be made reusable when autoclaved.
Ordering information: Available with flat or baffled base for increased agitation and aeration.
Customers Who Bought This Also Bought