VWR® Premium Plus Slide Boxes, 100-Place
Catalog #: 76265-652
Supplier: VWR International
Specifications
- Capacity:100-Place
- Color:Blue
- Description:VWR® Premium Plus Slide Box
- Cat. no.:76265-652
Specifications
About this item
Modern design for maximum functionality.
- Slide rail design ensures a secure fit, allowing easy placement and removal
- Centrally positioned location numbering grid enables simple slide organization with easy viewing of location numbers
- Sliding lock provides secure storage with easily identified lock and unlock symbols
- 2 mm zip tie can be used for more secure storage
- Inventory card is removable and replaceable for trouble-free writing on flat surfaces
- ABS plastic construction
The VWR® premium plus slide box's modern design for daily use is stylish with a wave surface texture. The slide box is easy to hold, increasing safety of sample transport with an EVA foam-lined bottom to hold slides securely in place and protect against damage. Unique side storage can accommodate a pen, desiccant, or other small items needed for working.
BodyTextAccessories: Removable and replaceable cardboard inventory card is available separately.
Customers Who Bought This Also Bought