Order Entry
ContactUsLinkComponent
Septum
DURAN® Silicone Seal, PTFE Coated (VMQ), DWK Life Sciences
Catalog #: 76264-276
Septum
DURAN® Silicone Seal, PTFE Coated (VMQ), DWK Life Sciences
Catalog #: 76264-276

Specifications

  • Ø:
    43 mm
  • Material:
    Silicone
  • Cat. no.:
    76264-276

Specifications

About this item

These seals are suitable for PBT screw caps and tamper-evident screw caps.

  • Autoclavable
  • Peroxidically-cured silicon
  • Heat resistance 130 °C (vapor) and 200 °C (dry heat)

Good chemical resistance due to PTFE coating.

Customers Who Bought This Also Bought