Plastic Spatulas, United Scientific Supplies
Catalog #: 76263-282
Supplier: United Scientific Supplies
Specifications
- Length:6"
- Description:Polypropylene flat and spoon spatula
- Cat. no.:76263-282
Specifications
About this item
Molded in polypropylene, spatulas feature one flat end and one spoon end.
- Polypropylene
- Autoclavable
This molded spoon is features an ergonomic deigned which make sit easy to hold.
Customers Who Bought This Also Bought