Utility Funnels, United Scientific Supplies
Catalog #: 76263-228
Supplier: United Scientific Supplies
Specifications
- Capacity:—
- Description:Set of Three (1 of each Size)
- Height:—
- Material:PP
- Top I.D.:—
- Stem Length:—
- Stem Diameter:—
Specifications
About this item
Useful for adding powder chemical to reaction vessels.
- Polypropylene
- Autoclavable
These funnels have eight internal ribs to facilitate filtration. They are made of polypropylene.
Customers Who Bought This Also Bought