Measuring Cylinders with Hexagonal Base, United Scientific Supplies
Catalog #: 76263-190
Supplier: United Scientific Supplies
Specifications
- Capacity:250 ml
- Graduation Interval:2 ml
- Graduation range:10 - 250 ml
- O.D.:46 mm
- Height:321 mm
- Cat. no.:76263-190
Specifications
About this item
Unbreakable polypropylene construction.
- Stable hexagonal base
- Unbreakable Polypropylene
Polypropylene one-piece measuring cylinders have raised graduations for easy readability. They are autoclavable and have excellent contact clarity. The hexagonal base provides excellent stability, and the tapered spout enables easy pouring of reagents. Stated tolerances are the same as graduated intervals below.
Customers Who Bought This Also Bought