UniStore™ Reagent Bottles, Narrow Mouth, HDPE
Catalog #: 76263-120
Supplier: United Scientific Supplies
Specifications
- Cap Size:20 mm
- Capacity:60 ml
- Height:85 mm
- Material:HDPE
- Cat. no.:76263-120
Specifications
About this item
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
- Translucent
- Pliable
UniStore™ narrow mouth HDPE reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.
Customers Who Bought This Also Bought