VWR® Cap Mat
Catalog #: 76221-326
Supplier: VWR International
Specifications
- Sterility:Non sterile
- Description:96-Well Round Well Cap Mat
- Material:EVA
- Cat. no.:76221-326
Specifications
About this item
Re-usable cap mats are available for friction sealing of plates.
Cap mats are an economic alternative to thermal or adhesive seals as they can be sterilized and reused.
Customers Who Bought This Also Bought