SiliaSep* Flash Cartridge 220g
Catalog #: CA76220-988
Supplier: SiliCycle
Customers Who Bought This Also Bought
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$72.65 to $846.50
VACUETTE® Blood Collection Tubes, without Additive, Greiner bio-one
Supplier: Greiner Bio-One
$13.18 to $316.03
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$500.19 to $1,790.36