BioClean-D™ Non sterile Coveralls with Hood and Integrated Boots, Ansell
Catalog #: CA75972-618
Supplier: Ansell Healthcare
Specifications
- Anti-static:Yes
- Clothing Size:S
- Color:White
- Seam type:Bound
- Cat. no.:CA75972-618
Specifications
About this item
The BioClean-D™ coverall with hood and integrated boots is a disposable garment offering comfort and head-to-toe protection.
- Anti-static, lightweight, low-linting CleanTough™ material construction
- Thumb loops to ensure a secure hold
- Slip resistant soles
- Excellent ESD properties
- Latex-free
Coveralls feature a front zip with protective flap and sealable cover, elasticated hood, back, cuffs, ankles, and thumb loops to ensure a secure hold. The integrated boots feature slip-resistant polyurethane soles to ensure every step can be taken with confidence.
Certifications: ASTM F739, EN1149-5, Category III.
Customers Who Bought This Also Bought