Order Entry
ContactUsLinkComponent
GENOMIC DNA KIT YEASTAR SPIN COL 40 PREP
YeaStar™ Genomic DNA Kit, Zymoresearch
Catalog #: 76212-304
Supplier:  Zymo Research
CAS Number:  
GENOMIC DNA KIT YEASTAR SPIN COL 40 PREP
YeaStar™ Genomic DNA Kit, Zymoresearch
Catalog #: 76212-304
Supplier:  Zymo Research
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Yeast/Fungi
  • Target molecule:
    DNA
  • Description:
    YeaStar™ Genomic DNA Kit
  • Size:
    40 preparations
  • Description:
    YeaStar™ Genomic DNA Kit
  • Cat. no.:
    76212-304

Specifications

About this item

Simple DNA purification from a broad spectrum of fungal species.

  • Efficient DNA isolation from a broad spectrum of fungal species susceptible to yeast lytic enzyme lysis
  • Genomic DNA can be used for southern blotting, PCR, restriction enzyme digestion
  • Equipment: Microcentrifuge, heat block/bath
  • Purity: Typical A260/A280 ≥1.8
  • Yield: Up to 25 µg total DNA can be eluted into ≥60 µl
  • Applicable for: All sensitive downstream applications such as qPCR and next-generation sequencing

The YeaStar™ Genomic DNA Kit is designed for reliable and efficient isolation of genomic DNA from a broad spectrum of fungal species, including A. fumigatus, A. nidulans, A. nivens var. aureus, C. albicans, pastoris, S. cerevisiae, S. pombe, and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis. The kit is based on highly efficient enzyme lysis and Zymo-Spin™ column technology. Each standard prep yields about 7 to 20 µg of DNA with a size distribution of 35 to 60 kb.

Caution: Stored at ≤–20 °C
Customers Who Bought This Also Bought