ContactUsLinkComponent
ZYMOTAQ DNA POLYMERASE HOT-START 200RXNS
ZymoTaq DNA Polymerase, Zymo Research
Catalog #: 76211-540
Supplier:  Zymo Research
CAS Number:  
ZymoTaq DNA Polymerase, Zymo Research
Catalog #: 76211-540
Supplier:  Zymo Research
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    ZymoTaq DNA Polymerase
  • Size:
    200 Reactions
  • Cat. no.:
    76211-540
Specifications
About this item
ZymoTaq DNA polymerase is a heat-activated, 'hot start' polymerase that has 3′-terminal transferase activity. The addition of 'A' overhangs to amplified DNA makes it ideal for use in TA-cloning.
  • Simple and easy to use
  • Compatible with real-time, quantitative PCR, and suitable for TA-cloning
  • Generates specific amplicons with little or no by-product formation
Reduces non-specific PCR product formation from difficult templates such as bisulfite converted DNA.
Customers Who Bought This Also Bought