Order Entry
ContactUsLinkComponent
VWR* Sleek Protective Eyewear, Blue Temple, Anti-Scratch, Anti-Fog, Clear Lens
VWR® Sleek Safety Glasses
Catalog #: 76210-826
Supplier:  VWR International
VWR* Sleek Protective Eyewear, Blue Temple, Anti-Scratch, Anti-Fog, Clear Lens
VWR® Sleek Safety Glasses
Catalog #: 76210-826
Supplier:  VWR International

Specifications

  • Lens Coating:
    Anti-fog, scratch-resistant
  • Lens Tint:
    Clear
  • Frame Color:
    Blue

Specifications

About this item

Lightweight and sleekly designed safety glasses.

  • Super lightweight at 22.7 g
  • Dielectric construction
  • Comfortable nose piece
  • Slim temple with rubber pad
  • Impact-resistant
  • Polycarbonate lens
  • Provides 99.9% UV protection
Certifications: Meets ANSI Z87.1 standards.
Customers Who Bought This Also Bought