VWR® OTG Safety Glasses
Catalog #: 76210-732
Supplier: VWR International
Specifications
- Lens Coating:Anti-fog, scratch-resistant
- Lens Tint:Clear
- Frame Color:Navy
Specifications
About this item
Economical and compact, designed to fit over prescription glasses.
- Fits over most prescription glasses
- Adjustable temple length for varied facial contours
- Polycarbonate lens
- Provides 99.9% UV protection
The pantoscopic adjustment allows lens angle modification and it is impact-resistant.
Certifications: Meets ANSI Z87.1 standards.
Customers Who Bought This Also Bought