Order Entry
ContactUsLinkComponent
Macrocentrifuge Tube with Snap Cap 5mL
Centrifuge Tubes with Snap Caps, 5 ml, Globe Scientific
Catalog #: 76209-496
Supplier:  Globe Scientific
Macrocentrifuge Tube with Snap Cap 5mL
Centrifuge Tubes with Snap Caps, 5 ml, Globe Scientific
Catalog #: 76209-496
Supplier:  Globe Scientific

Specifications

  • Sterility:
    Non sterile
  • Color:
    Black
  • Cat. no.:
    76209-496
  • Packaging:
    50/Bag

Specifications

About this item

These superior quality 5 ml centrifuge tubes with snap caps are an ideal alternate to 15 ml centrifuge tubes when working with smaller sample volumes.

  • Produced from polypropylene (PP)
  • Provided convenience of 1.5 ml microcentrifuge tube in a tube that holds larger volumes
  • Attached snap cap facilitates one-handed opening and closing
  • Pierceable cap allows easy access by needle or syringe
  • Dual graduation scales in increments of 0.5 and 0.2 ml enable more precise measurement when required
  • Suitable for use from –80 to 121 °C for low temperature storage and general laboratory use
  • Seven colors options for sample identification and organization

The 5 ml centrifuge tubes provide the convenience of a 1.5 ml microcentrifuge tube that hold larger volumes and are suitable for low temperature sample storage as well as general laboratory use.

Customers Who Bought This Also Bought