Oakton® PCTS Testr™ 50 Waterproof Pocket pH/Cond/TDS/Salinity Tester, Premium 50 Series, Cole-Parmer
Catalog #: 76208-092
Supplier: Argos Technologies
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Calibration:Up to 3 points
- Description:Waterproof Pocket pH/Cond/TDS/Salinity Tester
- Temperature accuracy:0 – 50 °C (±0.5 °C/±0.9 °F); 50 – 60 °C (±1.0 °C/±1.8 °F)
- Temperature resolution:0.1 °C (0.1 °F)
- Temperature range:0 – 60 °C (32 – 140 °F)
- Battery life:150 hrs.
- Automatic buffer recognition:USA 4.01, 7.00, 10.01 and NIST 4.01, 6.86, 9.18
- Conductivity accuracy:±1% full scale
- Conductivity range:0 µS - 20 mS
- Conductivity resolution:0.1 μS, 1 μS, 0.01 mS
- Salinity accuracy:±1% full scale
- Salinity range:0.00 – 10.0 ppt
- Salinity resolution:0.01 ppt
- TDS accuracy:±1% full scale
- TDS range:0 ppm – 10.0 ppt
- TDS resolution:10 ppt
- Temperature compensation:ATC
- pH accuracy:±0.01 pH
- pH range:–1 to 15
- pH resolution:0.01 pH
- Battery type:Four AAA 1.5 V
- Cat. no.:76208-092
Specifications
About this item
Ultimate tester for pH, conductivity, TDS, salinity, and temperature!
- Combine measurement capabilities of three testers in one convenient pocket-sized unit
- Up to 3 point pH calibration with NIST or USA auto buffer recognition
- Conductivity, TDS, and salinity feature auto ranging based on the concentration of your sample
- Waterproof
- LCD display
Redesigned leakproof cap functions as a sample/calibration cup, hydrating cup, and stabilizing base to stand the meter upright.
Delivery: Includes protective plastic storage case, lanyard, and 4 AAA batteries.
Customers Who Bought This Also Bought