Q-sep® QuEChERS Extraction Salts, Restek
Catalog #: 76204-760
Supplier: Restek
CAS Number:
Specifications
- Description:Q-sep® QuEChERS Extraction Salt Packets Only (Original)
- Size:50 Packets
- Cat. no.:76204-760
- Contents:4 g MgSO4, 1 g NaCl
Specifications
About this item
QuEChERS methods are fast, easy, and cost-effective, and Restek Q-sep® products make QuEChERS procedures even simpler.
- Free-flowing salts transfer easily and completely
- Easy-open packets eliminate the need for a second empty tube for salt transfer
- Convenient slim packets fit perfectly into tubes to prevent spills
- Ready-to-use tubes; no glassware required
- Pre-weighed, ultra-pure extraction salts
- Ideal for original unbuffered, AOAC (2007.01), and European (EN 15662) QuEChERS methods
No specialized glassware is required when using Q-sep® extraction packets and tubes. Free-flowing extraction salts and salt packets fit easily into the extraction tubes to make transferring salts to sample mess-free and easy.
Customers Who Bought This Also Bought