AccuGreen™ High Sensitivity and Broad Range dsDNA Quantitation Kits, Biotium
Catalog #: 76204-710
Supplier: Biotium
CAS Number:
Specifications
- Description:AccuGreen™ Broad Range dsDNA quantitation kit
- Size:500 assays
- Ex/Em:500/530 nm
- Cat. no.:76204-710
Specifications
About this item
The AccuGreen™ High Sensitivity and Broad Range dsDNA quantitation kits are designed for use with handheld fluorometers such as the Qubit® fluorometers from Thermo Fisher Scientific.
- Designed specifically for Thermo’s Qubit® fluorometer
- Linear range: 0.1 to 100 ng dsDNA (AccuGreen™ High Sensitivity), 2 to 1000 ng dsDNA (AccuGreen™ Broad Range)
- Sample concentration range: 5 pg/µl to 100 ng/µl (AccuGreen™ High Sensitivity), 0.1 to 1000 ng/µl (AccuGreen™ Broad Range)
- Green fluorescence emission
- Replaces Qubit® dsDNA HS and BR assay kits for cost savings
Fluorescent dsDNA quantitation kits for sensitive and selective detection of purified dsDNA samples on handheld fluorometers such as the Qubit® fluorometer. Kits available with or without DNA standards, for users that wish to use their own standards.
Customers Who Bought This Also Bought