Acetic acid glacial 99.5-100.05% USP, Multi-Compendial, J.T.Baker®
Catalog #: CAJT95263
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 64-19-7
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, clear
- Size:2.5 L
- Cat. no.:CAJT95263
Specifications
Specification Test Results
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- Meets J.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets B.P. Chemical Specifications:
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Identification:Passes Test
- USP - Congealing Temperature:≥15.6 °C
- USP - Nonvolatile Residue (USP), mg/20 mL:≤1.0
- USP - Chloride (Cl):Passes Test
- USP - Sulfate (SO₄):Passes Test
- USP - Readily Oxidizable Substances:Passes Test
- USP - Assay (CH₃COOH):99.5 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance:Passes Test
- EP/BP - Freezing Point:≥14.8 °C
- EP/BP - Iron (Fe):≤5 ppm
- EP/BP - Chloride (Cl):≤25 ppm
- EP/BP - Sulfate (SO₄):≤50 ppm
- EP/BP - Reducing Substances:Passes Test
- EP/BP - Residue on Evaporation:≤0.01 %
- EP/BP - Assay (CH₃COOH):99.0 - 100.5 %
- JP - Assay (CH₃COOH):99.0 - 101.0 %
- JP - Identification:Passes Test
- JP - Congealing Point:≥14.5 °C
- JP - Nonvolatile Residue, mg/10 mL:≤1.0
- JP - Chloride (Cl):Passes Test
- JP - Sulfate (SO₄):Passes Test
- JP - Heavy Metals (as Pb):≤10 ppm
- JP - Reducing Substances:Passes Test
- Residual Solvents: No Class 1, 2 or 3 residual solvents are used in or:
- produced by the production process.:
- Preserve in tight containers, and store at room temperature:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Acetic acid glacial 99.5-100.05% USP, Multi-Compendial:
Specification Test Results
About this item
Synonyms: Vinegar acid,Methanecarboxylic acid
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5-100,05
:Management of Change (MOC) category = R
Flash Pt:38.5 °C
:Boiling Pt:
:Boiling Pt: 118 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt: 38.5 °C
:M.W. 60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
REACH:01-2119475328-30
:8
:II
:GHS02
:GHS05
:GHS07
Customers Who Bought This Also Bought