Order Entry
ContactUsLinkComponent
Q tubes and caps, For Q qPCR instrument
Q qPCR Instrument, Quantabio
Catalog #: CA76202-252
Supplier:  Quantabio
Q qPCR Instrument, Quantabio
Catalog #: CA76202-252
Supplier:  Quantabio
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Q tubes and caps, 20 racks/ box
  • Cat. no.:
    CA76202-252

Specifications

About this item

A faster, smaller, better way to qPCR

  • Ultra-Fast Data Acquisition - 35 cycles in 25 minutes
  • Unrivaled Performance - Detect 2-fold expression level differences
  • Portable & Compact - 4.5 lbs - transport without ever calibrating
  • Scalable & Wireless - Connect up to 10 instruments (48 samples/instrument)
  • Magnetic Induction Technology - Eliminate variability vs block-based cyclers

Q uses a patented magnetic induction technology to rapidly heat samples coupled with fan forced air for cooling to acquire data in only 25 minutes. Available in 4 channel model, the robust optical system acquires all channels simultaneously and allows for running the fastest multiplexed assays.

Q’s miniature speaker-size and 4.5 pound weight make it the most portable and versatile qPCR cycler on the market without ever needing to calibrate. Q also provides scalability as each instrument can process up to 48 samples per run and up to 10 Q’s can be connected to a single computer wirelessly via bluetooth enabling up to 480 samples to be processed simultaneously.

A key difference is that Q incorporates a unique spinning aluminum rotor providing superior temperature uniformity of ± 0.05°C versus traditional block-based real time cyclers which rely on multiple peltier heating blocks that can create edge effects resulting in sample variation. Not only does the data give you superior reproducibility, repeatability but enables detection of 2-fold gene expression level differences as well as identification of difficult class IV SNP’s requiring melt temperature resolutions of 0.1°C.

Who wouldn’t want to take one for a spin?

Ultra-Fast Data Acquisition
Generate high quality data, fast!

Q’s speed is the fastest in the industry
Don’t sacrifice on the performance quality of your qPCR.
Completing runs in as little as 25 minutes* is the new standard

*25 minute cycle times obtained with fast cycling master mixes and short amplicon assay designs targeting cDNA

Unrivaled Performance
Detect 2-fold Differences

Confidently detect small differences.
High thermal uniformity and reproducibility
Detect differences within a single cycle

Wide Linear Dynamic Range
Ultra-sensitive detection down to single digit copies of DNA
Q performs reliably across a wide dynamic range

Q is the most portable and compact cyler on the market occupying ¼ the size of current qPCR platforms. The small footprint is enabled by the unique rotary and magnetic induction technology.

Fixed optics & no moving parts.
Never needs optical alignment or calibration
No reference dyes or cross talk compensation required
Utilizes proprietary 0.1 ml 4-strip tubes & caps supporting volumes of 5 – 30 μl
High speed centrifugation ensures sample spin down
Prevents evaporation & condensation with pre-loaded oil in tubes

Scalable & Wireless
Flexibility from 48 to 480 samples

Each Q cycler can process up to 48 samples per run and up to 10 cyclers can be connected to a single computer wirelessly to provide the desired scalability.

Combine single data sets from multiple runs from multiple cyclers
Data from different runs and cyclers look a like
Q is plug-and-play

Customers Who Bought This Also Bought