FlashPure Select C18 catridge 220g
Catalog #: 76196-952
Supplier: Buchi
Customers Who Bought This Also Bought
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
175,32 $ to 1 620,42 $
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
885,46 $ to 3 541,82 $
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
667,83 $
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier: MilliporeSigma
178,29 $
WEBCOL® Alcohol Preps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
15,59 $ to 18,12 $