FlashPure Select C18 catridge 80g
Catalog #: 76196-948
Supplier: Buchi
Customers Who Bought This Also Bought
Sodium thiosulfate 0.10 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$153.23 to $602.78
Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
$411.94 to $1,303.38
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$500.19 to $1,790.36