FlashPure Select C18 catridge 40g
Catalog #: 76196-946
Supplier: Buchi
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$73.73 to $459.25
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
$60.20 to $3,424.99
Quality Economy Brand Luer Lock Syringes with Attached Needle, Air-Tite
Supplier: Air-Tite
$40.13 to $365.76
Iodine monochloride solution acc. Wijs USP Test Solution (TS) for iodine absorption number of fats and oils
Supplier: Ricca Chemical
$210.52