FlashPure Select Silica catridge 12g
Catalog #: 76196-926
Supplier: Buchi
Customers Who Bought This Also Bought
Hydrion® Single Roll Dispenser, pH 1.0 to 12.0, Micro Essential Laboratory®
Supplier: Micro Essential
137,62 $
Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
411,94 $ to 1 303,38 $
Silver, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Supplier: VWR International
110,90 $
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
635,20 $ to 2 541,50 $
NAB Nanosep® Centrifugal Devices for RNA and DNA Extraction, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
141,42 $