Superpack Pipette Tips, MP Biomedicals
Catalog #: IC0977988H2
Supplier: MP Biomedicals
Specifications
- Volume:5 - 300 µl
- Packed:10×96/Box
- Sterile:+
Specifications
About this item
Specifically designed for use with the Titertek® and Labsystems Finnpipette®.
Tips may also be used with Biohit Proline™, Jencons Sealpette™, Labpette™, Nichiryo, Oxford-Benchmate™ and Pipetman® P-200.
Customers Who Bought This Also Bought