Disposable Micropipette Tips
Catalog #: 470233-654
Supplier: Edvotek
Specifications
- Volume:1000 - 5000 µl
- Packed:100 Tips/Bag
- Cat. no.:470233-654
Specifications
About this item
High quality micropipette tips to meet any lab's needs.
Quantities of 96 are supplied in a rack for easy dispensing; quantities of 1,000 are supplied in a plastic bag.
- Certified DNase, RNase and pyrogen free
- Material: PP
- Autoclavable
- Designed to fit most standard micropipettes
All quantities are supplied in bulk bags.
Customers Who Bought This Also Bought