Order Entry
ContactUsLinkComponent
Wards* pH Buffer Kit
Ward's® pH Buffer Kit
Catalog #: 470233-572
Supplier:  Ward's Science
CAS Number:  
Wards* pH Buffer Kit
Ward's® pH Buffer Kit
Catalog #: 470233-572
Supplier:  Ward's Science
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • pH:
    4/7/10
  • Description:
    Ward's Chemistry pH Buffer Kit
  • Size:
    Kit

Specifications

About this item

Includes 500 ml each of buffer solution pH 10, pH 4, and pH 7. Also includes 100 ml of electrode storage solution, KCl.

  • Aligned with GLOBE protocols

Shelf life: If stored in unopened bottles, then it is one year from receipt. Once opened it should be used within six months of first use.

Customers Who Bought This Also Bought