DuPont™ ProShield® 50 Coveralls with Respirator Hood
Catalog #: 76187-476
Supplier: DuPont
Specifications
- Clothing Size:L
- Color:White
- Cat. no.:76187-476
- Chest Size:251/4"
- Inseam:271/2"
- Sleeve:331/4"
Specifications
About this item
ProShield® 50 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
- Respirator fit hood
- Elastic wrists and ankles
- Storm flap
- Serged seams
ProShield® 50 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include: janitorial, sanitation, and general industrial maintenance.
Certifications: Thickness meets ASTM D751.
Customers Who Bought This Also Bought