Order Entry
ContactUsLinkComponent
Glove Dispenser Single Box White Steel
White Steel Glove Dispenser, ZING Enterprises
Catalog #: 76185-828
Supplier:  ZING Enterprises
Glove Dispenser Single Box White Steel
White Steel Glove Dispenser, ZING Enterprises
Catalog #: 76185-828
Supplier:  ZING Enterprises

Specifications

  • Dimensions:
    11.5 L x 4" W
  • Description:
    Glove Dispenser, Single Box
  • Material:
    Steel
  • Cat. no.:
    76185-828

Specifications

About this item

High quality dispensers are available in Single, Double, or Triple sizes.

  • Made from six gauge steel wire
  • Durable powder coat white finish
  • Mounting hardware included
  • Three sizes: Single, Double, and Triple
Customers Who Bought This Also Bought