Diamond™ Culture Tube Caps, Globe Scientific
Catalog #: 76184-764
Supplier: Globe Scientific
Specifications
- Color:Green
- Description:Diamond™ Culture Tube Caps
- Cat. no.:76184-764
- For Tube O.D.:13 mm
Specifications
About this item
These caps fit plain-end culture tubes and flasks with deLong style necks or ports.
- Available in five colors for easy sample identification and organization
- Molded from durable, autoclavable polypropylene (PP)
- Ridges inside cap provide secure grip and allow for gas exchange
- Maintain sterility without cotton plugging
- Reduce sample evaporation compared to cotton plugging
- Provides the same evaporation and sterility benefits as stainless steel closures
Customers Who Bought This Also Bought