Syringe Tips, Corning® Step-R™
12.5 ml, +, Individually Wrapped
Catalog # 76181-478
Supplier #: 6633
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
VWR TIP SYRINGE PIPETTOR STERILE 12.5ML Catalog #76163-610
Product 76163-610 is similar to 76181-478 but manufacturer and specifications may vary.
Product Details & Documents
Syringe tips for use with the Step-R™ repeating pipettor. Ideal for viscous, dense and volatile liquides.
- Minimum increments printed on the syringe itself
- Accurately deliver selected dose
- Made from high quality polypropylene and polyethylene
Available sterile or non sterile.
Ordering information: Tip adapters are required for using the 25 mL and 50 mL tips.
Specifications
- Volume:12.5 ml
- Packed:Individually Wrapped
- Sterile:+
Accessories
Product Information
- Catalog #VolumeAvailabilityPrice
- Corning® Step-R™ Repeating Adapter for 50 ml Syringe TipsCorning® Step-R™ Repeating Adapter for 50 ml Syringe Tips76193-27450 mlLog in for availabilityCase (50ml) - $8.23
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



