PIPETMAN® Expert Tipack™ Racked Tips, Gilson®
Catalog #: 76180-440
Supplier: GILSON, INC.
Product available on GSA Advantage®
Specifications
- Volume:0.1 - 20 µl
- Packed:3072 Tips
- Tip Style:Sterilized Tipack™ EL10ST
Specifications
About this item
One tip, any pipettor. Manufactured in a sterile, cleanroom environment using the purest raw materials and state-of-the-art techniques, users can be assured they will receive a high-quality product offering consistent performance.
- Ready to use
- Expert Tips are easy to pick up directly from the rack
- Tipack™ racks are fully autoclavable and sealed with a security sticker
- Racks are color-coded to differentiate tip models and indicate the compatible Gilson® pipettor model
To help prevent contamination, the 96 tips are loaded without manual intervention. Tipack™ racks are sealed with a security sticker and are fully autoclavable. They will resist being autoclaved several times. Using PIPETMAN® Tips is the only way to ensure accuracy and reproducibility across the full volume range of PIPETMAN® pipettors.
Packaging: Supplied in convenient and steady Tipack™ racks, come as 10 packs of 96 tips.
Customers Who Bought This Also Bought