PIPETMAN® Diamond Tips STERILPACK, Gilson®
Catalog #: 76180-032
Supplier: GILSON, INC.
Product available on GSA Advantage®
Specifications
- Volume:2 - 200 µl
- Packed:400 Tips Individually Wrapped
- Tip Style:D200 Diamond STERILPACK Tips
Specifications
About this item
Highest quality Diamond Tips for superior results.
Diamond Tips STERILPACKs are wrapped in medical device quality plastic, which provides a complete shield against contaminants. They are individually wrapped for complete protection up to the moment of use. Manufactured by a fully automatic process in a controlled environment, Diamond Tips are never touched by human hands.
Sterilized Diamond Tips are certified free of detectable DNA, RNA, DNase, RNase, ATP, pyrogens, and trace metals. Diamond Tips STERILPACKs are irradiated by gamma rays for more efficient sterilization. The sterilization is guaranteed for 5 years.
Autoclaving conditions: Moist heat/121°C/20 minutes/1 bar.
Customers Who Bought This Also Bought