PIPETMAN® Diamond Tips RELOAD Pack, Gilson®
Catalog #: 76178-588
Supplier: GILSON, INC.
Product available on GSA Advantage®
Specifications
- Volume:—
- Packed:—
- Cat. no.:76178-588
- Tip Style:Empty Box
- For Use With:D1000 RELOAD Pack
Specifications
About this item
Highest quality Diamond Tips for superior results.
The RELOAD Pack is the easiest and quickest way to refill the empty Tipack™ Box. Three times smaller than a regular tip box, this packaging is easy to store on all benches. Using up to 70% less plastic than a regular rack box, this new packaging is also eco-friendly. The racks are color-coded to differenciate tip models and indicate the compatible Gilson pipette models.
Autoclaving conditions: Moist heat/121 °C/20 min/1 bar.
Customers Who Bought This Also Bought