ContactUsLinkComponent
PIPETMAN* L manual pipette 20-300uL
PIPETMAN® L Multichannel Pipettor, Gilson®
Catalog #: 76178-370
Supplier:  GILSON, INC.
Product available on GSA Advantage®
PIPETMAN* L manual pipette 20-300uL
PIPETMAN® L Multichannel Pipettor, Gilson®
Catalog #: 76178-370
Supplier:  GILSON, INC.
Product available on GSA Advantage®
Specifications
  • Volume:
    20 - 300 µl
  • Description:
    Model P8x300L
  • For Use With:
    D200, D300, DF200 and DF300 Diamond Tips
  • No. of Channels:
    8
Specifications
About this item
PIPETMAN® L pipettes are enhanced from the legendary PIPETMAN Classic with a newly designed body for improved comfort and a state-of-the-art volume-locking mechanism for precision. These air-displacement pipettes are available in 10 multichannel models - volume ranges of 0.5 to 1200 µl.
  • Optimized comfort
  • Optimized safety and precision
  • Optimized traceability
PIPETMAN L pipettes, one of the most comfortable models in the Gilson portfolio, offer an ideal weight and handle shape. The natural fit of the pipette lets you relax your grip to make pipetting less fatiguing for your hand. To decrease the risk of repetitive strain injuries, the PIPETMAN L features an updated piston assembly with more responsive springs, which reduces pipetting forces. The tip ejector is also adjustable for right- or left-handedness, whichever is most comfortable for you.
PIPETMAN L offers volume-locking to ensure no accidental volume change during pipetting cycles, which could alter the accuracy and precision of your results. The patented locking system fully disconnects the operating rod from the counter assembly during pipetting so that once locked into place, the volume cannot be accidentally changed, preventing accidental volume drift during pipetting tasks. To help you minimize cross-contamination, variable volume models of PIPETMAN L are fully autoclavable with no disassembly required for maximum convenience.
PIPETMAN L features a 2D identification bar code for easy traceability and is particularly suited for enhanced GLP compliance. Using a universal scanner you can download the product part number, date of manufacturing and serial number.
The PIPETMAN L also has a color-coded push button that corresponds in color to Gilson PIPETMAN Tip packages.
Customers Who Bought This Also Bought