PIPETMAN Diamond Tips Empty Racks, Gilson®
Catalog #: 76178-272
Supplier: GILSON, INC.
Product available on GSA Advantage®
Specifications
- Description:Diamond Tips, Rack, Empty, D5000 Tips
- Cat. no.:76178-272
- For Use With:500-5000 µL PIPETMAN EXPERT Tips
Specifications
About this item
These empty racks can be used for ECOPACK and EASYPACK loose tips.
Customers Who Bought This Also Bought