Order Entry
ContactUsLinkComponent
PIPETMAN* TOWERPACK* Diamond Tip 100 - 1000uL
PIPETMAN® Diamond Tips TowerPack™, Gilson®
Catalog #: 76178-238
Supplier:  GILSON, INC.
Product available on GSA Advantage®
PIPETMAN* TOWERPACK* Diamond Tip 100 - 1000uL
PIPETMAN® Diamond Tips TowerPack™, Gilson®
Catalog #: 76178-238
Supplier:  GILSON, INC.
Product available on GSA Advantage®

Specifications

  • Volume:
    100 - 1000 µL
  • Packed:
    10 Tipack™ Racks of 96 Tips
  • Cat. no.:
    76178-238
  • Tip Style:
    D1000 Diamond TowerPack™

Specifications

About this item

Highest quality Diamond Tips for superior results.

The TowerPack™ is an innovative, quick, and easy refill system for PIPETMAN® Tips. Like all Gilson PIPETMAN® disposable pipette tips, the TowerPack™ is available in a sterilized version. Each sterilized TowerPack™ is individually wrapped in medical device quality plastic to help protect tips from contamination. Sterilization is guaranteed by a red sticker, which indicates exposure to gamma radiation. Save space and money, as the TowerPack™ uses 60% less space on a laboratory bench and 70% less plastic. One reload box accommodates the whole range of PIPETMAN® Tips with this easy-to-use, one-hand system. Save 15% on select sizes of TowerPack™ versus Tipack™ racks.

Autoclaving conditions: Moist heat/121 °C/20 min/1 bar.

Customers Who Bought This Also Bought