MICROMAN® Capillary Pistons, Gilson®
Catalog #: 76178-162
Supplier: GILSON, INC.
Product available on GSA Advantage®
Specifications
- Volume:50 - 250 µl
- Packed:960 Pistons
- Cat. no.:76178-162
- Tip Style:Capillary Pistons CP250
- For Use With:MICROMAN® M250E
Specifications
About this item
Capillary Pistons are designed, developed, and manufactured to fit the MICROMAN® perfectly.
- Made of pure PP
- Slim fit allows you to use the narrowest and deepest test tubes
Designed to fit the Gilson MICROMAN positive displacement pipette, Capillary Piston Tips are ideal for difficult-to-pipette samples including; viscous, volatile, dense, hot or cold, or hazardous liquids. The tips work like a syringe providing protection against pipette, sample, and user contamination, and produce accurate, reproducible, and precise results.
Certifications: ISO 8655-2.
Customers Who Bought This Also Bought