PIPETMAN® Single Channel Pipettor, Gilson®
Catalog #: 76178-044
Supplier: GILSON, INC.
Product available on GSA Advantage®
Specifications
- Volume:1 - 10 ml
- Description:PIPETMAN® P10 ml
- For Use With:D10 ml diamond tips
Specifications
About this item
Fully adjustable air-displacement pipettor PIPETMAN®.
- Extremely low pipetting forces
- Ergonomic and easy to use with premium comfort and performance
PIPETMAN® brings a new level of comfort into Gilson range with a redesigned piston mechanism that reduces the force needed to aspirate and dispense liquids. PIPETMAN® maintains the trusted durability, accuracy, and reliability of the original PIPETMAN® pipettes with comfort.
Certifications: Gilson maximum permissible errors are guaranteed only when PIPETMAN® pipettors are used with the recommended PIPETMAN® Diamond Tips.
Customers Who Bought This Also Bought