Specifications
- Light source:Xenon flash
- Description:GENESYS™ 50 UV-Visible Spectrophotometer
- Model:GENESYS™ 50, Dual Beam
- Electrical:100 - 240 V, 50 - 60 Hz
- Dimensions:355L×385W×195H mm
- Photometric accuracy:±0.002 A at 0.5 A;
±0.004 A at 1.0 A;
±0.008 A at 2.0 A - Photometric range:–2 to +3.5 A
- Repeatability:Photometric: ±0.001 A at 1 A (Measured at 1.0 A at 546 nm)
- Stray light:< 1.0% T 198 nm (KCl) , <0.05% T at 220 nm (NaI), <0.03% T at 340 nm (NaNO₂)
- Wavelength repeatability:<±0.2 nm
- Weight:7.5 kg
- Cat. no.:76176-740
- Light Type:Xenon flash lamp (>5 years typical, 3 years guaranteed)
- Wavelength:190 – 1100 nm
- Bandwidth:2 nm
Specifications
About this item
Built on the foundation of 60 years of spectroscopy experience, the next generation Thermo Scientific GENESYS family of Vis/UV-Vis spectrophotometers provides exceptional performance with a contemporary design.
- Start taking measurements immediately with the simple-to-use, high-resolution, 7-inch color-display touchscreen start menu
- Exchange samples with ease with large, accessible sample compartment
- Save and print your data fast any way you want it with multiple data-export options
- GENESYS 140, 150, 180 and BioMate 160 are Wi-Fi ready with optional USB adaptor
- Two year standard warranty with 1 year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
The Biomate 160 UV-Vis spectrophotometer includes Life Science Application Software package. Software includes pre-programmed life science methods such as Nucleic Acid A260; Protein A280 and A205; Protein Colorimetric assays and OD600.
The GENESYS 180 UV-Vis spectrophotometer incudes double beam optics. It is an ideal solution when a reference beam is required, such as for kinetics. Includes an automated 8-cell changer as standard.
There is a GENESYS spectrophotometer you can count on to provide accurate, reproducible results for your application requirements.